Learn More
Lenz Laborglasintrumente Burette Stopcock, Lateral, with PTFE Plug
Product Code. 10131471
Change view
Click to view available options
:
NS 12.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10131471 NS 12.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10131471 Supplier Lenz Laborglasintrumente Supplier No. 02702501
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Facetted tip
  • Stopcock: NS 12.5
  • Bore: 1.5 mm
  • NS PTFE plug
  • Screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Material Borosilicate Glass (3.3)
Stopcock NS 12.5