Learn More
Abeba™ Business Lady 3990 Shoes
Comfortable with smooth leather upper. Antistatic.
Brand:  Abeba™ 3990/38
Additional Details : Weight : 0.66500kg
 View more versions of this product
Product Code. 15631288
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Smooth leather
  • Black
  • Antistatic leather outsole sole
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Heel 60 mm
Specifications
Specifications
Black
Leather
Leather
Microfiber
38
3990