missing translation for 'onlineSavingsMsg'
Learn More

Buspirone.HCl, Lipomed

Product Code. 30460954
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460954 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460954 Supplier Lipomed Supplier No. LPMBSP1832HC100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Buspirone.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buspirone hydrochloride
CAS 33386-08-2
Analyte or Component Names Buspirone hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H31 N5 O2 . Cl H
Synonym Buspirone Hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5 ]decane-7,9-dione;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 421.96
Quantity 100 mg
Formula Weight 421.2245 g/mol
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.