missing translation for 'onlineSavingsMsg'
Learn More

Buspirone hydrochloride, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60476.06

Quantity 5g

 View more versions of this product

Product Code. 15403249

  • 195.00€ / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

Solubility
Soluble in methanol (50 mg/ml), water (partly), DMSO (100 mM).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

33386-08-2
421.97
RICLFGYGYQXUFH-UHFFFAOYSA-N
36431
8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
C21H32ClN5O2
MFCD00078569
buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
CHEBI:3224
C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl

Specifications

33386-08-2
MFCD00078569
UN2811
buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
RICLFGYGYQXUFH-UHFFFAOYSA-N
8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
36431
421.96
C21H32ClN5O2
5g
14,1504
Soluble in methanol (50mg/ml),water (partly),DMSO (100 mM).
C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl
421.97
CHEBI:3224
Buspirone hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.