missing translation for 'onlineSavingsMsg'
Learn More

Buspirone hydrochloride, Thermo Scientific™

59.20€ - 195.00€

Chemical Identifiers

CAS 33386-08-2
Molecular Formula C21H32ClN5O2
Molecular Weight (g/mol) 421.97
MDL Number MFCD00078569
InChI Key RICLFGYGYQXUFH-UHFFFAOYSA-N
Synonym buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
PubChem CID 36431
ChEBI CHEBI:3224
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
SMILES C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493239
SDS View Documents
Thermo Scientific Alfa Aesar
J60476.03
1g
59.20€
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15403249
SDS View Documents
Thermo Scientific Alfa Aesar
J60476.06
5g
195.00€
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

Solubility
Soluble in methanol (50 mg/ml), water (partly), DMSO (100 mM).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

33386-08-2
421.97
RICLFGYGYQXUFH-UHFFFAOYSA-N
36431
8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
C21H32ClN5O2
MFCD00078569
buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
CHEBI:3224
C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl

Specifications

33386-08-2
MFCD00078569
14,1504
Soluble in methanol (50mg/ml),water (partly),DMSO (100 mM).
C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl
421.97
CHEBI:3224
Buspirone hydrochloride
C21H32ClN5O2
UN2811
buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
RICLFGYGYQXUFH-UHFFFAOYSA-N
8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
36431
421.96
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 251-489-4

missing translation for 'rtecsNumber' : CL9915000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Buspirone hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.