missing translation for 'onlineSavingsMsg'
Learn More

Buspirone hydrochloride, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60476.03


Quantity 1g

 View more versions of this product

Product Code. 15493239

  • 59.20€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is widely used in cell signaling and neuroscience research. Buspirone has been used to mitigate the electrically stimulated 5-hydroxytryptamine outflow from rat cortical slices.

Solubility
Soluble in methanol (50 mg/ml), water (partly), DMSO (100 mM).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

33386-08-2
421.97
RICLFGYGYQXUFH-UHFFFAOYSA-N
36431
8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
C21H32ClN5O2
MFCD00078569
buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
CHEBI:3224
C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl

Specifications

33386-08-2
MFCD00078569
UN2811
buspirone hydrochloride, buspar, buspirone hcl, narol, buspimen, ansitec, anxinil, anxiolan, busirone, buspinol
RICLFGYGYQXUFH-UHFFFAOYSA-N
8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
36431
421.96
C21H32ClN5O2
1g
14,1504
Soluble in methanol (50mg/ml),water (partly),DMSO (100 mM).
C1CCC2(C1)CC(=O)N(C(=O)C2)CCCCN3CCN(CC3)C4=NC=CC=N4.Cl
421.97
CHEBI:3224
Buspirone hydrochloride
Suggestions de produits

Suggestions de produits

Vidéos
Sécurité et traitement

Sécurité et traitement

Identifiant produit
  • Buspirone hydrochloride

Mention d'avertissement
  • Danger

Catégorie de danger
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Mention de danger
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

FDS Documents

Documents

Product Certifications
Promotions

Promotions

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.