missing translation for 'onlineSavingsMsg'
Learn More

Buspirone, TRC

Product Code. 30137034
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137034 1 g 1g
30133727 250 mg 250mg
18062250 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137034 Supplier Toronto Research Chemicals Supplier No. TRCB6898551G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36505-84-7
Molecular Formula C21 H31 N5 O2
Molecular Weight (g/mol) 385.5
Synonym 8-Azaspiro[4.5]decane-7,9-dione, 8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]- (9CI, ACI), 8-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione (ACI), Buspin, Buspirone, MeSH ID: D002065, Spitomin
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione
SMILES O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H31N5O2/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20/h5,8-9H,1-4,6-7,10-17H2
Formula Weight 385.2478
Percent Purity >95%
Chemical Name or Material Buspirone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.