missing translation for 'onlineSavingsMsg'
Learn More

Butadiene Monoxide 97%, ACROS Organics™

Product Code. 12744663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle packed in Plastic container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12744663 5g; Glass bottle packed in Plastic container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12744663 Supplier Thermo Scientific Acros Supplier No. 155790050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 930-22-3
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle in Plastic Container
Molecular Formula C4H6O
Refractive Index 1.4160 to 1.4200 (20°C, 589nm)
MDL Number MFCD00005149
Quantity 5 g
Beilstein 17, I, 13
Fieser 04,53
Synonym butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide
InChI Key GXBYFVGCMPJVJX-UHFFFAOYSA-N
SMILES C=CC1CO1
IUPAC Name 2-ethenyloxirane
Molecular Weight (g/mol) 70.091
PubChem CID 13586
Formula Weight 70.09
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Butadiene monoxide, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.