missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acetate, ACS Reagent, ≥99.5%, Honeywell™ Riedel-de Haën™

Product Code. p-7120266
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 15685400

Brand: Honeywell-Riedel-de Haen 402842500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Melting Point -78°C
Density 0.88g/mL at 25°C
Boiling Point 126.2°C (1013 hPa)
Explosion Limit 0.076
Flash Point 23°C (73.4°F)
Ignition Point 790°F
Odor Like fruit
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Quantity 500 mL
UN Number UN1123
Beilstein 1741921
Vapor Density 4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 8mmHg (20°C), 15mmHg (25°C)
Percent Purity ≥99.5%
Chemical Name or Material Butyl acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.