missing translation for 'onlineSavingsMsg'
Learn More

Butyl Benzoate, TRC

Product Code. 30347907
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347907 500 mg 500mg
30344872 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30347907 Supplier Toronto Research Chemicals Supplier No. TRCB120000500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyl benzoate
CAS 136-60-7
Molecular Formula C11 H14 O2
Quantity 500 mg
Synonym Benzoic acid n-butyl ester, Butyl benzoate, Chemcryl C 101N, IP Carrier N 20, NSC 8474, n-Butyl benzoate
InChI Formula InChI=1S/C11H14O2/c1-2-3-9-13-11(12)10-7-5-4-6-8-10/h4-8H,2-3,9H2,1H3
SMILES CCCCOC(=O)c1ccccc1
IUPAC Name butyl benzoate
Molecular Weight (g/mol) 178.23
Formula Weight 178.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.