missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Carbamate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Butyl Carbamate |
| CAS | 592-35-8 |
| Molecular Formula | C5H11NO2 |
| Quantity | 1 g |
| Synonym | Butyl carbamate, NSC 187, n-Butyl carbamate, O-Butyl carbamate |
| InChI Formula | InChI=1S/C5H11NO2/c1-2-3-4-8-5(6)7/h2-4H2,1H3,(H2,6,7) |
| SMILES | CCCCOC(=O)N |
| IUPAC Name | butyl carbamate |
| Molecular Weight (g/mol) | 117.15 |
| Formula Weight | 117.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction