missing translation for 'onlineSavingsMsg'
Learn More

Butyl Methacrylate, Mikromol™

Product Code. 17934006
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17934006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17934006 Supplier Mikromol Supplier No. MM0753.01
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butyl Methacrylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methacrylic acid butyl ester
CAS 97-88-1
Analyte or Component Names Methacrylic acid butyl ester
Shipping Condition Room Temperature
Molecular Formula C8 H14 O2
Certifications/Compliance ISO 9001
Synonym Basic Butylated Methacrylate Copolymer Imp. A (EP), Methacrylic acid, butyl ester (6CI,8CI), 2-Methyl-2-propenoic acid butyl ester, Acryester B, Acryester BMA, Blemmer BMA, Butyl 2-methyl-2-propenoate, Butyl 2-methylacrylate, Butyl methacrylate, Light Ester NB, N 212, NSC 20956, n-Butyl methacrylate, Basic Butylated Methacrylate Copolymer Impurity A
InChI Formula InChI=1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3
SMILES CCCCOC(=O)C(=C)C
IUPAC Name butyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 142.2
Quantity 100 mg
Formula Weight 142.0994 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.