missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyric-4,4,4-d3 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Butanoic Acid 4,4,4-D3 |
| CAS | 36789-14-7 |
| Molecular Formula | C4H5D3O2 |
| Quantity | 0.25 g |
| Synonym | Butanoic-4,4,4-d3 acid (9 CI), Butyric-4,4,4-d3 acid, Butanoic acid 4,4,4-D3 |
| InChI Formula | InChI=1 S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3 |
| SMILES | OC(CCC([2 H])([2 H])[2 H])=O |
| IUPAC Name | butanoic-4,4,4-d3 acid |
| Molecular Weight (g/mol) | 91.12 |
| Formula Weight | 91.07 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction