missing translation for 'onlineSavingsMsg'
Learn More

Butyryl Chloride, TRC

Product Code. 30330518
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330518 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330518 Supplier Toronto Research Chemicals Supplier No. TRCB8105105G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyryl Chloride
CAS 141-75-3
Molecular Formula C4H7ClO
Quantity 5 g
Synonym Butyryl Chloride, Butanoic Acid Chloride, Butyric Acid Chloride, Butyric Chloride, Butyroyl Chloride, n-Butanoyl Chloride, n-Butyroyl Chloride, n-Butyryl Chloride
InChI Formula InChI=1S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3
SMILES CCCC(=O)Cl
IUPAC Name butanoyl chloride
Molecular Weight (g/mol) 106.55
Formula Weight 106.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.