missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyryl-d7 Chloride, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Butyryl-d7 Chloride |
| CAS | 1219805-71-6 |
| Molecular Formula | C4 D7 Cl O |
| Quantity | 1 g |
| Synonym | Butanoyl-2,2,3,3,4,4,4-d7 chloride (ACI), 2,2,3,3,4,4,4-Heptadeuteriobutanoyl chloride, Butyryl-d7 Chloride, Butanoyl Chloride-D7, Butanoyl chloride-d7 |
| InChI Formula | InChI=1 S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)Cl |
| IUPAC Name | 2,2,3,3,4,4,4-heptadeuteriobutanoyl chloride |
| Molecular Weight (g/mol) | 113.594 |
| Formula Weight | 113.062 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction