missing translation for 'onlineSavingsMsg'
Learn More

[c]-Benzocarbazole, TRC

Product Code. 30355691
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355691 50 mg 50mg
30363862 100 mg 100mg
30354616 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355691 Supplier Toronto Research Chemicals Supplier No. TRCC98747350MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205-25-4
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.27
Synonym 7H-Benzo(C)Carbazole
IUPAC Name 7H-benzo[c]carbazole
SMILES c1ccc2c(c1)ccc3[nH]c4ccccc4c23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H11N/c1-2-6-12-11(5-1)9-10-15-16(12)13-7-3-4-8-14(13)17-15/h1-10,17H
Formula Weight 217.0891
Chemical Name or Material [c]-Benzocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.