missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C. botulinum BoNT-D Heavy Chain Monoclonal Antibody (682810)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561696
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The BoNT-D Heavy Chain is a crucial component of botulinum neurotoxin type D (BoNT-D), which is produced by Clostridium botulinum, a gram-positive, spore-forming bacterium known for causing botulism. BoNT-D is one of the seven serotypes of botulinum toxin, each of which is highly potent and capable of causing severe paralytic illness. The heavy chain of BoNT-D plays a vital role in the neurotoxin's mechanism of action. It is responsible for binding to specific receptors on the surface of nerve cells and facilitating the internalization of the toxin into the cell. Once inside, the heavy chain aids in the translocation of the light chain into the cytoplasm, where the light chain exerts its enzymatic activity by cleaving SNARE proteins, thereby inhibiting neurotransmitter release and leading to flaccid paralysis. Understanding the structure and function of the BoNT-D heavy chain is essential for the development of antitoxins and therapeutic interventions. This knowledge is also crucial for advancing medical applications of botulinum toxins in treating conditions such as muscle spasticity, chronic pain, and other disorders related to neurotransmission. Research into the heavy chain of BoNT-D not only helps in managing botulism but also expands the potential therapeutic uses of botulinum toxins.
Specifications
| C. botulinum BoNT-D Heavy Chain | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| E. coli-derived recombinant Clostridium botulinum Neurotoxin Type D Heavy Chain, Asn862-Glu1276. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 682810 | |
| Unconjugated | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction