Learn More
RuvA Antibody (RuvA 12C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50278-0.025ml
 View more versions of this product
Product Code. 18677247
  • 190.00€ / 0.03mL
Description
RuvA Monoclonal antibody specifically detects RuvA in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RuvA
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
EC 3.1.22.4, Holliday junction ATP dependent DNA helicase ruvA, Ruv A
Ruv A protein from E. Coli
0.025 mL
Epigenetics
E. coli
Purified
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
RuvA 12C6
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.