missing translation for 'onlineSavingsMsg'
Learn More

C.I.Solvent Orange 3, TRC

Product Code. 30136371
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136371 10 g 10g
30135962 5 g 5g
17950705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136371 Supplier Toronto Research Chemicals Supplier No. TRCC22434510G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-54-5
Molecular Formula C12 H12 N4
Molecular Weight (g/mol) 212.25
Synonym 1,3-Benzenediamine, 4-(phenylazo)- (9CI), C.I. Solvent Orange 3 (8CI), 4-(2-Phenyldiazenyl)-1,3-benzenediamine, 2,4-Diaminoazobenzene, 4-(Phenylazo)-1,3-phenylenediamine, Azobenzene-2,4-diamine, C.I. 11270:1, Chrysoidine Base, Chrysoidine Base A, Chrysoidine Base B, Chrysoidine G Base, Chrysoidine J Base, Chrysoidine Y Base, Chrysoidine Y Base New, Chrysoidine YD Base, Fat Brown GG, Grasan Chrysoidine, NSC 3273, Neptune Orange Base 206, Solvent Orange 3, Waxoline Orange Y, 1,3-Benzenediamine, 4-(2-phenyldiazenyl)-
IUPAC Name 4-[(E)-phenyldiazenyl]benzene-1,3-diamine
SMILES Nc1ccc(N=Nc2ccccc2)c(N)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H12N4/c13-9-6-7-12(11(14)8-9)16-15-10-4-2-1-3-5-10/h1-8H,13-14H2
Formula Weight 212.1062
Chemical Name or Material Solvent Orange 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.