missing translation for 'onlineSavingsMsg'
Learn More

C.I.Vat Yellow 2 (Technical Grade), TRC

Product Code. 30179484
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179484 500 mg 500mg
30179451 250 mg 250mg
30178941 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179484 Supplier Toronto Research Chemicals Supplier No. TRCC475860500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-09-9
Molecular Formula C28H14N2O2S2
Molecular Weight (g/mol) 474.56
IUPAC Name 6,16-diphenyl-5,15-dithia-7,17-diazapentacyclo[11.7.0.03,11.04,8.014,18]icosa-1(13),3(11),4(8),6,9,14(18),16,19-octaene-2,12-dione
SMILES O=C1c2ccc3nc(sc3c2C(=O)c4ccc5nc(sc5c14)c6ccccc6)c7ccccc7

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C28H14N2O2S2/c31-23-18-12-14-20-26(34-28(30-20)16-9-5-2-6-10-16)22(18)24(32)17-11-13-19-25(21(17)23)33-27(29-19)15-7-3-1-4-8-15/h1-14H
Formula Weight 474.0497
Chemical Name or Material C.I.Vat Yellow 2 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.