missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ c-Raf Recombinant Rabbit Monoclonal Antibody (JJ086-06)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA541270

Product Code. 17451423

  • 502.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

c-Raf Recombinant Monoclonal Antibody for Western Blot

Raf-1 (c-Raf) is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated Raf-1 can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylate to activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

c-Raf
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P04049
Raf1
Synthetic peptide within human Raf1 aa 1-50/648.
100 μL
Primary
Human
Antibody
IgG
Western Blot
JJ086-06
Unconjugated
Raf1
6430402F14Rik; AA990557; BB129353; CMD1NN; CRAF; C-RAF; C-Raf proto-oncogene, serine/threonine kinase; Craf1; Craf1 transforming; D830050J10Rik; EC 2.7.11.1; kinase Raf1; murine leukemia viral (v-raf-1) oncogene homolog 1 (3611-MSV); murine sarcoma 3611 oncogene 1; NS5; Oncogene RAF1; OTTHUMP00000207813; OTTHUMP00000209389; proto-oncogene c-RAF; RAF; raf proto-oncogene serine/threonine protein kinase; RAF proto-oncogene serine/threonine-protein kinase; RAF1; Raf-1; Raf-1 proto-oncogene, serine/threonine kinase; sarcoma 3611 oncogene; v-Raf; v-raf-1 leukemia viral oncogene 1; vraf1 murine le; v-raf-1 murine leukemia viral oncogene homolog 1; v-raf-1 murine leukemia viral oncogene-like protein 1; v-raf-leukemia viral oncogene 1
Rabbit
Protein A
RUO
5894
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.