missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | C11orf70 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16810355
|
Proteintech
20780-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16800355
|
Proteintech
20780-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| C11orf70 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q9BRQ4 | |
| 85016 | |
| C11orf70 Fusion Protein Ag14793 | |
| Primary | |
| -20°C | |
| C11orf70 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CK070, C11orf70 | |
| C11orf70 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title