Learn More
C15orf27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26094-1-AP-20UL
 View more versions of this product
Product Code. 16804625
  • 145.00€ / 20µL
Specifications
C15orf27
Polyclonal
Unconjugated
TMEM266
C15orf27, Transmembrane protein C15orf27
Rabbit
Antigen Affinity Chromatography
RUO
123591, 244886
-20°C
Liquid
Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q2M3C6, Q8BZB3
Tmem266
C15orf27 Fusion Protein Ag23390
20 μL
Primary
Human, Mouse