missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ C1orf54 (1-131) 293T Cell Transient Overexpression Lysate (Denatured)

Product Code. 16114283
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16114283 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16114283 Supplier Abnova™ Supplier No. H00079630T05.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human overexpression lysate

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description C1orf54 (1-131) 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC017761
Molecular Weight (g/mol) Theoretical M.W. 14.63kDa
Plasmid pCMV-C1orf54 (1-131) partial fragment
Quality Control Testing Transient overexpression cell lysate was tested with anti-C1orf54 (1-131) antibody (Mfr. No. H00079630-B04) by Western Blot.
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.