missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24474-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes one of two duplicated zinc finger genes on chromosome Xp11. This gene is the telomeric copy; GeneID 158586 ZXDB is the more centromeric copy. The two genes have 98% nucleotide sequence similarity, and the predicted proteins contain 10 tandem zinc finger motifs.Specifications
| C1QBP | |
| Polyclonal | |
| Unconjugated | |
| C1QBP | |
| C1QBP, gC1Q R, GC1q R protein, GC1QBP, gC1qR, Glycoprotein gC1qBP, HABP1, Hyaluro binding protein 1, p32, p33, SF2p32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12261, 708 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35658, Q07021 | |
| C1QBP | |
| C1QBP Fusion Protein Ag19773 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction