missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1QTNF9 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149219
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Probable adipokine. Activates AMPK, AKT, and p44/42 MAPK signaling pathways.
Specifications
| C1QTNF9 | |
| Polyclonal | |
| Unconjugated | |
| C1QTNF9 | |
| 9130217G22Rik; AQL1; C1q and tumor necrosis factor related protein 9; C1QTNF9; C1QTNF9A; Ciqtnf9; Complement C1q and tumor necrosis factor-related protein 9; complement C1q and tumor necrosis factor-related protein 9A; complement C1q tumor necrosis factor-related protein 9; complement C1q tumor necrosis factor-related protein 9A; CTRP9; RGD1560876; UNQ6503/PRO21380 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 338872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P0C862 | |
| C1QTNF9 | |
| Mouse myeloma cell line NS0-derived recombinant human CTRP9/C1qTNF9, Asn16-Pro333. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction