missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf149 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19912-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| C20orf149 | |
| Polyclonal | |
| Unconjugated | |
| PPDPF | |
| C20orf149, EXPDF, PPDPF, PPDPF,EXPDF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66496, 79144 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CR37, Q9H3Y8 | |
| Ppdpf | |
| C20orf149 Fusion Protein Ag13740 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction