missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24678-1-AP-150UL
This item is not returnable.
View return policy
Description
C20orf46, also named as TMEM74B, belongs to the TMEM74 family.Specifications
| C20orf46 | |
| Polyclonal | |
| Unconjugated | |
| TMEM74B | |
| C20orf46, Transmembrane protein C20orf46 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108832, 55321 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NUR3 | |
| TMEM74B | |
| C20orf46 Fusion Protein Ag20385 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction