missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28025-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| C2orf56 | |
| Polyclonal | |
| Unconjugated | |
| NDUFAF7 | |
| C2orf56, NDUFAF7, PRO1853 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298748, 55471, 73694 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XI79, Q7L592, Q9CWG8 | |
| NDUFAF7 | |
| C2orf56 Fusion Protein Ag27590 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction