missing translation for 'onlineSavingsMsg'
Learn More

uvex C300 Wet Plus Safety Gloves

Product Code. p-7230484 Shop All Uvex Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17240441 7 10 pairs
17250441 8 10 pairs
17260441 9 10 pairs
17270441 10 10 pairs
17280441 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17240441 Supplier uvex Supplier No. 6054607
Add to basket
Add to basket
This item is not returnable. View return policy

Bamboo TwinFlex™ Technology provides level C cut protection, while SoftGrip foam coating on the palm and fingertips enhances grip capabilities.

  • SoftGrip coating on the palm and fingertips for a high level of abrasion resistance
  • Bamboo TwinFlex™ Technology provides level C cut protection
  • Climazone™ ensures the highest possible user comfort
  • Enhanced grip is wet areas and oily working conditions
  • Free of harmful substances in accordance with Oeko-Tex™ Standard 100
  • Silicone-free according to imprint test
  • Resistant to oil and grease
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material HPE
Color Gray, Black
Hand Ambidextrous
Latex Free Yes
Material Bamboo, HPPE, Polyamide
Product Type Cut-resistant Gloves
Size 7
EN388 Standard 4X42C
For Use With (Application) Automotive, Maintenance, Metal Work, and Precision Assembly
Gender Unisex
Glove Cuff Style Knit
Series C300 Wet Plus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.