Learn More
C4orf36 Monoclonal Antibody (OTI8B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808287S
Description
C4orf36 Monoclonal antibody specifically detects C4orf36 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| C4orf36 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| C4orf36; chromosome 4 open reading frame 36; uncharacterized protein C4orf36 | |
| Full length human recombinant protein of human MGC12965 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8B7 | |
| Unconjugated | |
| LOC100287896 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100287896 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.