Learn More
C9orf41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24068-1-AP-150UL
 View more versions of this product
Product Code. 16872655
  • 410.00€ / 150µL
Description
N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).
Specifications
Specifications
C9orf41
Polyclonal
Unconjugated
CARNMT1
C9orf41, UPF0586 protein C9orf41
Rabbit
Antigen Affinity Chromatography
RUO
138199
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N4J0
CARNMT1
C9orf41 Fusion Protein Ag21268
150 μL