Learn More
C9orf41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24068-1-AP-150UL
This item is not returnable.
View return policy
Description
N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).Specifications
| C9orf41 | |
| Polyclonal | |
| Unconjugated | |
| CARNMT1 | |
| C9orf41, UPF0586 protein C9orf41 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 138199 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N4J0 | |
| CARNMT1 | |
| C9orf41 Fusion Protein Ag21268 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |