missing translation for 'onlineSavingsMsg'
Learn More

CA9 Mouse anti-Human, Mouse, Porcine, Clone: 1G4G5, Proteintech

Mouse Monoclonal Antibody

144.20€ - 410.97€

Specifications

Antigen CA9
Clone 1G4G5
Concentration 1.36 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887935
View Documents
Proteintech
66243-1-IG-20UL
20 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
16877935
View Documents
Proteintech
66243-1-IG-150UL
150 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Carbonic anhydrase (CA) is an enzyme that assists rapid interconversion of carbon dioxide and water into carbonic acid, protons, and bicarbonate ions. It is abundant in all mammalian tissues. There are many genes that are inducible by hypoxia, via HIF-1 alpha. CA IX is one of the most inducible genes because of its stability and location within the membrane. Carbonic anhydrases have a widespread role in regulating pH in normal tissues, by regulating hydrogen ion (H+) flux. The pH is important in cell death under hypoxia, thus a blockade of CA IX results in increased cell death under hypoxia. Therefore, CA IX has become a reliable histochemical marker of hypoxia.
Specifications

Specifications

CA9
1.36 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CA IX, CA9, CAIX, Carbonate dehydratase IX, Carbonic anhydrase 9, carbonic anhydrase IX, G250, Membrane antigen MN, MN, P54/58N, pMW1, RCC associated antigen G250
CA9
IgG2a
Protein G
Antibody
1G4G5
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
Mouse, Human, Pig
Q16790, Q8VHB5
100152792, 230099, 768
CA9 Fusion Protein Ag1540
Primary
-20°C
CA9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.