missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
390.00€ - 535.00€
Specifications
Material | Glass |
---|---|
For Use With (Application) | For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar |
Sterility | Non-sterile |
Product Code | Brand | Needle Gauge | Includes | Length (English) Needle | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Includes | Length (English) Needle | Product Type | Price | Quantity & Availability | ||
15886084
|
Cadence Science™
7977 |
18 G | Reinforcing bar | 2.87 in. | Micro Emulsifying Needle with Reinforcing Bar |
535.00€
Pack of 4 |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11769275
|
Cadence Science™
7978 |
20 G | Without reinforcing bar | 1.87 in. | Micro Emulsifying Needle w/o Reinforcing Bar |
390.00€
Pack of 4 |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
- Available with or without a reinforcing bar
- Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)
Specifications
Glass | |
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar | |
Non-sterile |