missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 463.00€
Specifications
| Antigen | Cadherin-9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647211
|
Novus Biologicals
NBP2-92182-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627030
|
Novus Biologicals
NBP2-92182-0.1ml |
0.1 mL |
463.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-9 Polyclonal antibody specifically detects Cadherin-9 in Human samples. It is validated for Western BlotSpecifications
| Cadherin-9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1007 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cadherin 9, type 2 (T1-cadherin) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 340-420 of human CDH9 (NP_057363.3). QMLYTLRVDASNTHPDPRFLHLGPFKDTAVVKISVEDIDEPPVFTKVSYLIEVDEDVKEGSIIGQVTAYDPDARNNLIKYS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title