missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcein, Reagent grade, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Specifications
Specifications
| Formula Weight | 622.5 |
| Grade | Research |
| Color | Orange |
| Quantity | 10 g |
| Physical Form | Powder |
| Chemical Name or Material | Calcein |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction