accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1461-15-0

HOONHOOOOHNOHOOHOOOHO

CAS RN 1461-15-0

IUPAC Name: 2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid
Synonyms: calcein oftasceina oftasceine oftasceinum 奥荧光素
Molecular Weight (g/mol): 622.54
Molecular Formula: C30H26N2O13
InChi Key: DEGAKNSWVGKMLS-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)CC1=CC2=C(OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C22OC(=O)C3=CC=CC=C23)C=C1O
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (3)
  • (3)
  • (2)
  • (4)
Color 
  • (2)
  • (7)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (3)
  • (9)
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (7)
  • (3)