missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate Monohydrate, ≥99%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11402441 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402441 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402441 Supplier MP Biomedicals Supplier No. 0219382190
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 5743-26-0
Quantity 500 g
Assay Percent Range ≥99%
Molecular Formula C4H8CaO5
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]
IUPAC Name calcium;diacetate;hydrate
Molecular Weight (g/mol) 176.181
PubChem CID 82163
ChEBI CHEBI:59199
Formula Weight 176.2
Grade ACS Reagent
Chemical Name or Material Calcium Acetate Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.