missing translation for 'onlineSavingsMsg'
Learn More

Calcium Arsenate, TRC

Product Code. 30345691
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345691 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345691 Supplier Toronto Research Chemicals Supplier No. TRCC1450151G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Arsenate
CAS 7778-44-1
Molecular Formula 2 As O4 . 3 Ca
Quantity 1 g
Synonym Calcium Arsenate, Chip-Cal Granular, Cucumber Dust, Pencal, Spra-cal, Tricalcium Arsenate
InChI Formula InChI=1S/2AsH3O4.3Ca/c2*2-1(3,4)5;;;/h2*(H3,2,3,4,5);;;/q;;3*+2/p-6
SMILES [Ca+2].[Ca+2].[Ca+2].[O-][As](=O)([O-])[O-].[O-][As](=O)([O-])[O-]
IUPAC Name tricalcium;trioxido(oxo)-λ^{5}-arsane
Molecular Weight (g/mol) 398.072
Formula Weight 397.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.