missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Dihydrate, MP Biomedicals™

Product Code. 11485092 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485092 5 kg 5kg
11466511 100 g 100g
11476511 500 g 500g
11430952 500 g 500g
11465092 500 g 500g
11475092 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11485092 Supplier MP Biomedicals Supplier No. 0219463505
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calcium chloride, Dihydrate Cell Culture Reagent, ∽99%, White Powder

  • Calcium chloride is a commonly used reagent in biochemistry.
  • It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
  • Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
  • It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
  • Calcium Chloride is a salt of calcium and chloride.
  • It behaves as a typical ionic halide, and is solid at room temperature

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613
Synonym Calcium dichloride dihydrate

Specifications

Melting Point 176°C (Decomposition)(Literature)
pH 4 to 9.5 (5% aq soln)
Physical Form Powder
Quantity 5 kg
Assay Percent Range ∽99%
Molecular Formula CaCl2·2H2O
Solubility Information Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.
Molecular Weight (g/mol) 147
Formula Weight 147
Chemical Name or Material Calcium Chloride Dihydrate
Grade ∽99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.