missing translation for 'onlineSavingsMsg'
Learn More

Calcium D-saccharate tetrahydrate, 98%

Product Code. 15438519
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15438519

Brand: Thermo Scientific Alfa Aesar J66947.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5793-89-5 | C6H16CaO12 | 320.26 g/mol

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.

Solubility
Soluble in water(0.43g/L).

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5793-89-5
Molecular Formula C6H16CaO12
Molecular Weight (g/mol) 320.26
MDL Number MFCD00150704
InChI Key ZQWFSIZRQANUDA-WQMSYZFBSA-L
Synonym calcium saccharate, unii-6ap9j91k4v, d-saccharic acid calcium salt tetrahydrate, calcium d-glucarate tetrahydrate, calcium d-saccharate tetrahydrate, calcium saccharat-4-wasser, calcium saccharate usan:usp:inn, calcium saccharate usp, calcium d-glucarate, tetrahydrate, calcium d-glucarate 1:1 , tetrahydrate
PubChem CID 16211545
IUPAC Name calcium;(2R,3S,4S,5S)-2,3,4,5-tetrahydroxyhexanedioate;tetrahydrate
SMILES C(C(C(C(=O)[O-])O)O)(C(C(=O)[O-])O)O.O.O.O.O.[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Beilstein 3821334
Merck Index 14,1700
Solubility Information Soluble in water(0.43g/L).
Formula Weight 320.26
Percent Purity 98%
Odor Odorless
Chemical Name or Material Calcium D-saccharate tetrahydrate
compliance-icons
Product Identifier
  • Calcium D-saccharate tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : D-Glucaric acid, calcium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.