missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard: Ca @ 2000 μg/g in 75 cSt Hydrocarbon Oil

Product Code. 30310359
Change view
Click to view available options
Quantity:
200 g
Unit Size:
200g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310359 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310359 Supplier VHG Labs Supplier No. VHGOCA2000200G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium
CAS 7440-70-2
Analyte or Component Names Calcium
Shipping Condition Room Temperature
Molecular Formula Ca
Concentration 2000 μg/g
Synonym Ca
InChI Formula InChI=1 S/Ca
Solution Type Single Solution
SMILES [Ca]
Element Ca
IUPAC Name calcium
Solvent or Matrix 75 cSt Hydrocarbon Oil
Industry Type Industrial QC
Molecular Weight (g/mol) 40.078
Quantity 200 g
Formula Weight 39.963
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.