missing translation for 'onlineSavingsMsg'
Learn More

Calcium trifluoromethanesulfonate, 98%, ACROS Organics™

Product Code. 10613183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10613183 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10613183 Supplier Thermo Scientific Acros Supplier No. 358680100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 55120-75-7
Color Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C2CaF6O6S2
Linear Formula (F3CSO3)2Ca
MDL Number MFCD00143911
Quantity 10 g
Synonym calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate
InChI Key PUQLFUHLKNBKQQ-UHFFFAOYSA-L
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2]
IUPAC Name calcium;trifluoromethanesulfonate
Molecular Weight (g/mol) 338.204
PubChem CID 2734307
Formula Weight 338.21
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Calcium trifluoromethanesulfonate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.