Learn More
CALCR Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20868-1-AP-20UL
 View more versions of this product
Product Code. 16870445
  • 145.00€ / 20µL
Description
The CALCR antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CALCR. This antibody recognizes human, mouse antigen. The CALCR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CALCR
Polyclonal
Unconjugated
20 μL
799
IgG
Western Blot, ELISA
400 μg/mL