Learn More
CALCR Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20868-1-AP-20UL
Description
The CALCR antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CALCR. This antibody recognizes human, mouse antigen. The CALCR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CALCR | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 799 | |
| IgG |
| Western Blot, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |