missing translation for 'onlineSavingsMsg'
Learn More
Learn More
calcyphosine Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | calcyphosine |
|---|---|
| Clone | 2B7D1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237577
|
Proteintech
68225-1-IG-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235860
|
Proteintech
68225-1-IG-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The calcyphosine antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human calcyphosine. This antibody recognizes human, pig, rabbit antigen. The calcyphosine antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| calcyphosine | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 828 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2B7D1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Rabbit | |
| CAPS1; MGC126562 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title