Learn More
Unity Lab Services™ Calibration Report Bath
Product Code. 11759208
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration Unit Size
11759208 1 Day Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11759208 Supplier Unity Lab Services™ Supplier No. UMVP9312
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
  • Stability control at a temperature setpoint in the center of the bath
  • Setpoint in the range -35°C to +200°C
  • Fluids used: pure alcohol from -35°C to +5°C, water up to +90°C, and silicone oil up to +200°C
  • Reference instrument certified by a COFRAC accredited laboratory
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermostatted Baths
Duration 1 Day
Product Type Calibration Report