Learn More
Calreticulin Antibody (681233), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB38981MFV610
Product Code. 18542529
  • 684.00€ / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
Calreticulin Monoclonal antibody specifically detects Calreticulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calreticulin
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
811
Store at 4C in the dark.
Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
681233
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated
autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA
E. coli-derived recombinant human Calreticulin, Glu18-Leu417, Accession # P27797
RUO
Primary
Human, Mouse, Rat
IgG2b