missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (CL12842) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18545AF405
This item is not returnable.
View return policy
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Calretinin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| This antibody was developed using a recombinant protein derived from P22676, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Phospho Specific | |
| 794 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL12842 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction