missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Flexible method-choice of assay protocols
- Rapid assay protocol
- Non-radioactive
- Precise and accurate measurement
- Ready-to-use substrate
- Color coded reagents
- Five-hour protocol
- Kits available with or without acetylation reagents
- Bulk kit provided without lysis buffers
- Dual range: 25–6400 fmol/well [non-acetylation], 2–128 fmol/well [acetylation]
- Sensitivity: 38.4pg/mL [non-acetylation] and 14pg/mL [acetylation]
Specifications
Specifications
| Assay Sensitivity | 38.4 pg/mL (non-Ac), 14 pg/mL (Ac) |
| Product Type | cAMP biotrak Enzyme immunoassay (EIA) system |
| Sufficient For | 5 x 96-well Microplate |
| For Use With (Application) | Used for research purposes, eliminates lengthy sample extraction procedures and allow direct cAMP measurement |
| Kit Contents | 96-well coated plates, Assay buffer, Non-acetylation assay standard, Peroxidase conjugate antiserum, TMB substrate, Wash buffer |
| Quantity | 5 x 96 Determinations |
| Storage Requirements | 2°C to 8°C |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?