missing translation for 'onlineSavingsMsg'
Learn More

Campestanol, TRC

Product Code. 30011718
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011718 10 mg 10mg
30011627 5 mg 5mg
30003376 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011718 Supplier Toronto Research Chemicals Supplier No. TRCC15529010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474-60-2
Molecular Formula C28 H50 O
Molecular Weight (g/mol) 402.7
Synonym Ergostan-3-ol, (3β,5α,24R)-, 5α-Ergostan-3β-ol, (24R)- (8CI), Campestanol (7CI), (3β,5α,24R)-Ergostan-3-ol, 24β-Ethylcholestanol, 24β-Methylcholestanol, 5α-Dihydrocampesterol, Chalinastanol, Neospongosterol, dihydro-, Dihydrocampesterin, (24R)-3-ß-Hydroxy-24-methyl-5a-cholestane, (24R)-5α-Ergostan-3β-ol, Dihydro-neospongosterol, CAMP
IUPAC Name (3S,5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(1R,4R)-1,4,5-trimethylhexyl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)[C@H](C)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C28H50O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h18-26,29H,7-17H2,1-6H3/t19-,20-,21+,22+,23+,24-,25+,26+,27+,28-/m1/s1
Formula Weight 402.3862
Percent Purity >95%
Chemical Name or Material Campestanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.